Sign In to View Organizational & Contract Pricing.
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
biological source
rabbit
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
mouse
technique(s)
immunoprecipitation (IP): suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
mouse ... Rab32(67844)
General description
Ras-related protein Rab32 is a mitochondrion localized protein that acts as an anchoring protein for protein kinase A regulatory subunit which then plays a role in mitochondrial fission and mitochondrial dynamics of growth and recycling. Rab32 is also involved in phagosome maturation where it is recruited and serves as a lipid anchoring protein for various pathogens. Rab32 is expressed in my tissues in both the adult and the developing fetus. Rab32 is part of the Rab GTPase superfamily of GTP signaling proteins. Interestingly, some human specific pathogens such as Salmonella enterica need to replicate intracellularly and have evolved mechanisms to avoid Rab32/phagosome mediated engulfment and subsequent destruction. It appears to some species deliver an effector protein that cleaves Rab32 and thus are then free to develop intracellular vacuole-like vesicles in which to replicate and avoid destruction.
~25 kDa observed
Immunogen
GST-tagged recombinant protein corresponding to mouse Rab32.
Application
Research Category
Apoptosis & Cancer
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Apoptosis - Additional
This Anti-Rab32 Antibody is validated for use in Western Blotting and Immunoprecipitation for the detection of Rab32.
Western Blotting Analysis: A representative lot detected Rab32 in B16-f1 cell lysates (Ohbayashi, N., et al. (2012). MBoC. 23:669-678).
Western Blotting Analysis: A representative lot detected Rab32 in Melan-A cell lysates (Tamura, K., et al. (2009). MBoC. 20:2900-2908).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Rab32 in B16-f1 cell lysate (Ohbayashi, N., et al. (2012). MBoC. 23:669-678).
Western Blotting Analysis: A representative lot detected Rab32 in Melan-A cell lysates (Tamura, K., et al. (2009). MBoC. 20:2900-2908).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Rab32 in B16-f1 cell lysate (Ohbayashi, N., et al. (2012). MBoC. 23:669-678).
Physical form
Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified
Preparation Note
Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
Evaluated by Western Blotting in Melan-A cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Rab32 in 10 µg of Melan-A cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Rab32 in 10 µg of Melan-A cell lysate.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.