Skip to Content
Merck
All Photos(1)

Key Documents

AB9298

Sigma-Aldrich

Anti-MARCKS Antibody, Complete sequence

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MARCKS(4082)

Specificity

Recognizes MARCKS.

Immunogen

Full length recombinant rat MARCKS.

Application

Anti-MARCKS Antibody, Complete sequence is an antibody against MARCKS for use in WB.
Immunoblotting: 1:1,000 using rat brain extract (colormetric; higher dilutions may be used with ECL). The antibody reacts with the 70-85 kDa (depending on species) MARCKS protein. The suggested dilution buffer is TBS containing 0.05% Tween-20. Suggested blocking buffer is 5% non-fat dry milk. Suggested transfer membrane is PVDF-Immobilon. Suggested gel percentage is 8%. Overnight incubation with the antibody is recommended.

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: AB5427

Physical form

Format: Purified
Semi-purified immunoglobulin. Liquid in 120 mM NaCl, 25 mM Tris-HCl, pH 7.3 containing 0.01% sodium azide.

Analysis Note

Control
POSITIVE CONTROL: Neonatal rat brain extract. MARCKS is heat and acid resistant. Heat or acid-treated supernatants from crude tissue or cell homogenates are good sources to analyze MARCKS content by immunoblot.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Yu et al.
PloS one, 10(11), e0141397-e0141397 (2015-11-04)
Overexpression of the myristolated alanine-rich C kinase substrate (MARCKS) occurs in vascular proliferative diseases such as restenosis after bypass surgery. MARCKS knockdown results in arrest of vascular smooth muscle cell (VSMC) proliferation with little effect on endothelial cell (EC) proliferation.
Dan Yu et al.
Journal of vascular surgery, 70(6), 2021-2031 (2019-04-02)
Restenosis limits the durability of all cardiovascular reconstructions. Vascular smooth muscle cell (VSMC) proliferation drives this process, but an intact, functional endothelium is necessary for vessel patency. Current strategies to prevent restenosis employ antiproliferative agents that affect both VSMCs and
Dan Yu et al.
Journal of the American Heart Association, 4(10), e002255-e002255 (2015-10-10)
Transcription of the myristoylated alanine-rich C kinase substrate (MARCKS) is upregulated in animal models of intimal hyperplasia. MARCKS knockdown inhibits vascular smooth muscle cell (VSMC) migration in vitro; however, the mechanism is as yet unknown. We sought to elucidate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service