Recommended Products
biological source
rabbit
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
guinea pig, rat
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
immunohistochemistry: suitable
western blot: suitable
shipped in
dry ice
target post-translational modification
unmodified
Specificity
Glutamate
The cross-reactivities were determined using an ELISA test by competition experiments with the following compounds:
Compound Cross-reactivity
L-Glutamate-G-BSA 1
D-Glutamate-G-BSA 1/>50,000
L-Aspartate-G-BSA 1/>50,000
D-Aspartate-G-BSA 1/>50,000
GABA-G-BSA 1/>50,000
The antisera was also tested for specificity using the free-floating PAP technique on rat cortex.
Abbreviations:
(G)Glutaraldehyde
(=)Non-reduced conjugate
(BSA) Bovine Serum Albumin
The cross-reactivities were determined using an ELISA test by competition experiments with the following compounds:
Compound Cross-reactivity
L-Glutamate-G-BSA 1
D-Glutamate-G-BSA 1/>50,000
L-Aspartate-G-BSA 1/>50,000
D-Aspartate-G-BSA 1/>50,000
GABA-G-BSA 1/>50,000
The antisera was also tested for specificity using the free-floating PAP technique on rat cortex.
Abbreviations:
(G)Glutaraldehyde
(=)Non-reduced conjugate
(BSA) Bovine Serum Albumin
Immunogen
L-Glutamate-gluteraldehyde-BSA
Application
Detect Glutamate using this Anti-Glutamate Antibody validated for use in ELISA, IH & WB.
Research Category
Neuroscience
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Neuroinflammation & Pain
Neurotransmitters & Receptors
Neuroinflammation & Pain
Physical form
Rabbit antiserum. Liquid with 0.05% sodium azide.
Storage and Stability
Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.
During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.
During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Aaron E Rusheen et al.
Brain : a journal of neurology, 146(10), 4174-4190 (2023-05-04)
Tourette syndrome is a childhood-onset neuropsychiatric disorder characterized by intrusive motor and vocal tics that can lead to self-injury and deleterious mental health complications. While dysfunction in striatal dopamine neurotransmission has been proposed to underlie tic behaviour, evidence is scarce
Elenia Cinelli et al.
Neuroscience, 326, 126-140 (2016-04-09)
Neurons within the vagal motoneuron region of the lamprey have been shown to modulate respiratory activity via ascending excitatory projections to the paratrigeminal respiratory group (pTRG), the proposed respiratory rhythm generator. The present study was performed on in vitro brainstem
Hannah E Westlake et al.
Journal of morphology, 278(1), 29-49 (2016-10-04)
Staurozoan cnidarians display an unusual combination of polyp and medusa characteristics and their morphology may be informative about the evolutionary origin of medusae. We studied neuromuscular morphology of two staurozoans, Haliclystus 'sanjuanensis' and Manania handi, using whole mount immunohistochemistry with
Jesper Ericsson et al.
The Journal of physiology, 591(4), 859-874 (2012-11-14)
The striatum of the basal ganglia is conserved throughout the vertebrate phylum. Tracing studies in lamprey have shown that its afferent inputs are organized in a manner similar to that of mammals. The main inputs arise from the thalamus (Th)
Zhenyu Liu et al.
Journal of neurophysiology, 115(3), 1477-1486 (2016-01-15)
In many regions of the vertebrate brain, microcircuits generate local recurrent activity that aids in the processing and encoding of incoming afferent inputs. Local recurrent activity can amplify, filter, and temporally and spatially parse out incoming input. Determining how these
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service