Skip to Content
Merck
All Photos(1)

Documents

869019-M

Sigma-Aldrich

Normal Rabbit Serum, Lyophilized Solid

Synonym(s):

Normal Rabbit Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.41

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<0.05% Hemoglobin

storage temp.

−20°C

Related Categories

General description

Normal carrier sera, produced from pooled lots of sera from healthy, non-immunized rabbits. Suitable for use in radioimmunoassay.

Warning

Toxicity: Standard Handling (A)

Physical form

pH 6.5-8.0.

Reconstitution

Reconstitute in 5 ml of H₂O.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service