Skip to Content
Merck
All Photos(1)

Documents

8.41424

Sigma-Aldrich

tert-Butylhydroquinone

for synthesis

Synonym(s):

tert-Butylhydroquinone, 2-tert.-Butyl-1,4-dihydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
MDL number:
UNSPSC Code:
12352104
EC Index Number:
217-752-2
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

powder

autoignition temp.

457 °C

potency

700 mg/kg LD50, oral (Rat)

bp

295 °C/1013 hPa

mp

127-129 °C

transition temp

flash point 171 °C

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C10H14O2/c1-10(2,3)8-6-7(11)4-5-9(8)12/h4-6,11-12H,1-3H3

InChI key

BGNXCDMCOKJUMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 126 °C
Melting range (upper value): ≤ 129 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service