Skip to Content
Merck
All Photos(1)

Documents

8340-OP

Millipore

SSC Buffer

OmniPur® Grade, 20X, Premixed Powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319

Quality Level

form

fine powder (free-flowing)

manufacturer/tradename

Calbiochem®

color

white

shipped in

ambient

General description

Each pouch makes 1 L of 20X SSC buffer (1X =0.150 M Sodium Chloride, 0.015 M Sodium Citrate). Molecular biology grade.

Specifications:
Chloride content (1%, water): 110-120 meq/L
DNase: None detected
pH (1%, water) at 25°C: 7.85-8.15
Protease: None detected
RNase: None detected
Solubility (1.0%, water): To pass test





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service