Skip to Content
Merck
All Photos(1)

Documents

8.22068

Sigma-Aldrich

Hydroxyethylcellulose

for synthesis

Synonym(s):

Hydroxyethylcellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.22

Quality Level

form

granular

autoignition temp.

>360 °C

bulk density

350 kg/m3

storage temp.

2-30°C

InChI

1S/C29H52O21/c1-10-15(34)16(35)24(13(8-33)45-10)49-28-20(39)18(37)25(50-29-26(43-5-4-30)21(40)23(42-3)12(7-32)47-29)14(48-28)9-44-27-19(38)17(36)22(41-2)11(6-31)46-27/h10-40H,4-9H2,1-3H3

InChI key

CWSZBVAUYPTXTG-UHFFFAOYSA-N

Analysis Note

Assay (Cellulose ether): ≥ 92.0 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service