Skip to Content
Merck
All Photos(1)

Key Documents

8.14437

Sigma-Aldrich

Tributylacetyl citrate

for synthesis

Synonym(s):

Tributylacetyl citrate, 2-(Acetyloxy)-1,2,3-Propanetricarboxylic acid tributyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O8
CAS Number:
Molecular Weight:
402.48
MDL number:
UNSPSC Code:
12352108
EC Index Number:
201-067-0

vapor pressure

0.07 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (acidimetric)

form

liquid

bp

170-173 °C/1 hPa

mp

-80 °C

transition temp

flash point 204 °C

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C20H34O8/c1-5-8-11-25-17(22)14-20(28-16(4)21,19(24)27-13-10-7-3)15-18(23)26-12-9-6-2/h5-15H2,1-4H3

InChI key

QZCLKYGREBVARF-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 1.049 - 1.052
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

422.6 °F - closed cup

Flash Point(C)

217 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service