Skip to Content
Merck
All Photos(1)

Key Documents

71542

Sigma-Aldrich

Trx•Tag Monoclonal Antibody

Novagen®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

antibody form

purified antibody

Quality Level

antibody product type

primary antibodies

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Detection of Trx•Tag fusion proteins

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service