Skip to Content
Merck
All Photos(1)

Key Documents

6.17816

Sigma-Aldrich

Imidazole

purified grade

Synonym(s):

Imidazole

Sign Into View Organizational & Contract Pricing

Select a Size

IN26178160100
₪34.00

₪34.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
IN26178160100
₪34.00

About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
MDL number:
UNSPSC Code:
12352005

₪34.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

purified grade

SMILES string

[nH]1cncc1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service