Skip to Content
Merck
All Photos(1)

Key Documents

6.09065

Supelco

Perchloric acid solution

0.1 mol/l in anhydrous acetic acid, for titrations in non-aqueous media

Synonym(s):

Perchloric acid, PCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352301

Product Name

Perchloric acid, 0.1 mol/l in anhydrous acetic acid for titrations in non-aqueous media

grade

anhydrous

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application



  • Nanoporous gold with microporous structure: Perchloric acid is used in the electrochemical dealloying process to create nanoporous gold structures, which are significant for their applications in catalysis and sensor technology (Li et al., 2024).


  • Stability indicating reversed-phase-high-performance liquid chromatography method: This method uses perchloric acid for the analysis of pharmaceutical compounds, offering enhanced stability and reliability for drug formulation and safety checks (Chintala, 2024).


  • Development of innovative methodology for determination of 6-thioguanine: By utilizing perchloric acid, this HPLC-PDA-based technique allows for precise medical diagnostics, facilitating better disease management through accurate drug monitoring (Noga et al., 2023).


Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service