Skip to Content
Merck
All Photos(1)

Key Documents

585970

Sigma-Aldrich

(±)-Thalidomide

Selective inhibitor of TNF-α biosynthesis.

Synonym(s):

(±)-Thalidomide, (±)-2-(2,6-Dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N2O4
CAS Number:
Molecular Weight:
258.23
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C13H10N2O4/c16-10-6-5-9(11(17)14-10)15-12(18)7-3-1-2-4-8(7)13(15)19/h1-4,9H,5-6H2,(H,14,16,17)

InChI key

UEJJHQNACJXSKW-UHFFFAOYSA-N

General description

Selective inhibitor of TNFα biosynthesis. Blocks basic fibroblast growth factor (bFGF)-induced angiogenesis in cornea. An immunomodulator that inhibits replication of the HIV-1 virus. Has been used to treat AIDS-related oral ulcers.
Selective inhibitor of TNF-α biosynthesis. Blocks basic fibroblast growth factor (bFGF)-induced angiogenesis in cornea. An immunomodulator that inhibits replication of the HIV-1 virus. Has been used to treat AIDS-related oral ulcers.

Biochem/physiol Actions

Cell permeable: no
Primary Target
TNF-α biosynthesis
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Toxic & Carcinogenic / Teratogenic (G)

Reconstitution

Following reconstitution, aliquot and freeze(-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Gershon, D. 1995. Nat. Med.1, 1230.
Thompson, C. 1995. Lancet 346, 1289.
Tramontana, J.M., et al. 1995. Mol. Med. 1, 384.
D’Amato, R.J., et al. 1994. Proc. Natl. Acad. Sci. USA 91, 4082.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service