Skip to Content
Merck
All Photos(1)

Key Documents

5.30618

Sigma-Aldrich

Pin1 Inhibitor, DTM

Synonym(s):

Pin1 Inhibitor, DTM, PPIase-Parvulin Inhibitor II

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
₪915.00

₪915.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
₪915.00

About This Item

Empirical Formula (Hill Notation):
C12H20N2S3
CAS Number:
Molecular Weight:
288.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

₪915.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell permeable thiuram monosulfide based compound that acts as a competitive inhibitor of Pin1 (EC50 = 4.1 µM, Ki = 50 nM). Binds to the active site of the PPIase domain of Pin1 hydrophobically and blocks the active pocket. Shown to specifically inhibit peptidyl-prolyl cis/trans isomerase activity in HeLa cells. Inhibits the proliferation of HCT116 cells (EC50 = 10 µM) by arresting cell cycle at the G0 phase. Also shown to reduce the endogenous Runx2 protein levels in the MC3T3-E1 cells. Displays excellent solubility in DMSO (~100 mM).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Pin1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Yoon, W-J., et al. 2013. J. Cell Physiol.228, 2377.
Tatara, Y., et al. 2009.Biochem Biophys Res Commun.384, 394.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service