Skip to Content
Merck
All Photos(1)

Key Documents

5.05736

Sigma-Aldrich

5-BDBD

Synonym(s):

5-BDBD, 5-(3-Bromophenyl)-1,3-dihydro-2H-Benzofuro[3,2-e]-1,4-diazepin-2-one, P2X4 Purinergic Receptor Antagonist, 5-BDBD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11BrN2O2
CAS Number:
Molecular Weight:
355.19
MDL number:
UNSPSC Code:
12352200

Assay

≥99% (HPLC)

Quality Level

form

powder

potency

500 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 15 mg/mL

storage temp.

2-8°C

InChI

1S/C17H11BrN2O2/c18-11-5-3-4-10(8-11)15-17-16(20-14(21)9-19-15)12-6-1-2-7-13(12)22-17/h1-8H,9H2,(H,20,21)

InChI key

NKYMVQPXXTZHSF-UHFFFAOYSA-N

General description

A highly potent antagonist of purinergic P2X4 receptor. Shown to block P2X4-mediated currents in Chinese hamster ovary cells (IC50 = 500 nM). Blocks ATP-induced dilation of spiral ligament capillaries in guinea pigs. Also shown to restore the viability of activated CD4+ cells in the presence of 1 mM ATP.

Biochem/physiol Actions

Primary Target
P2X₄

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Trabanelli, S., et al. 2012. J. Immunol.189, 1303.
Wu, T., et al. 2011. Am. J. Physiol.301, H69.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service