Skip to Content
Merck
All Photos(1)

Key Documents

5.04521

Sigma-Aldrich

Hhat Inhibitor, RU-SKI 43

Synonym(s):

Hhat Inhibitor, RU-SKI 43, Hedgehog Acyltransferase Inhibitor, RU-SKI43, 2-(2-Methylbutylamino)-1-(4-(m-tolyloxymethyl)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)ethanone, 2-((2-Methylbutyl)amino)-1-(4-((3-methylphenoxy)methyl)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)ethanone, RU-SKI43, 2-(2-Methylbutylamino)-1-(4-(m-tolyloxymethyl)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)ethanone, 2-((2-Methylbutyl)amino)-1-(4-((3-methylphenoxy)methyl)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)ethanone, Hedgehog Acyltransferase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30N2O2S
CAS Number:
Molecular Weight:
386.55
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

semisolid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

InChI

1S/C22H30N2O2S/c1-4-16(2)13-23-14-22(25)24-10-8-21-19(9-11-27-21)20(24)15-26-18-7-5-6-17(3)12-18/h5-7,9,11-12,16,20,23H,4,8,10,13-15H2,1-3H3

InChI key

AEENEMOEBJOKGN-UHFFFAOYSA-N

General description

A cell-permeable dihydrothienopyridinyl-ethanone compound that acts as an effective inhibitor against MBOAT (membrane-bound O-acyltranferase) family member Hhat/hedgehog acyltransferase-catalyzed ShhN palmitoylation in cell-free assays (by 70% at 12.5 µM) in a palmitoyl CoA-noncompetitive (Ki = 6,9 µM; [ShhN] = 38 µM) and ShhN-uncompetitive (Ki = 7.4 µM; [palmitoyCoA] = 30 µM) manner. Effectively prevents Shh- & Hhat-transfected COS-1 cells from activating hedgehog signaling in co-cultured C3H10T1/2 cells (~80% inhibition of AP production in 30 min with10 µM inhibitor) by inhibiting COS-1 intracellular ShhN palmitoylation (by 52% and 80%, respectively, with 10 or 20 µM drug treatment and serum withdrawal 1 h before 4 h [125I]-palmitate incubation). RU-SKI 43 is shown not to interfere with cellular palmitoylation of H-ras & fyn, myristoylation of c-Src & Wnt3a, Shh autoprosessing, or the protein level and localization of Shh & Hhat. Exhibits short plasma half-life in mice (t1/2 = 17 min; i.v.).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Hhat
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Petrova, E., et al. 2013. Nat. Chem. Biol.9, 247.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service