Skip to Content
Merck
All Photos(1)

Documents

444231

Sigma-Aldrich

MMP-9, Monomer, Human Neutrophil

Synonym(s):

Matrix Metalloproteinase 9, Gelatinase B, 92 kDa Type IV Collagenase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥1000 mU/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

foreign activity

other MMP activity, none detected

shipped in

wet ice

storage temp.

−70°C

General description

Monomeric, native MMP-9 from stimulated human neutrophils. Matrix metalloproteinases (MMP) are members of a family of at least 20 zinc-dependent endoproteinases that function extracellularly at neutral pH. The enzymes are synthesized as latent or inactive zymogens that contain a regulatory propeptide. This propeptide confers latency by binding to the Zn2+ molecule present in the active site. MMP activation can be achieved through autoactivation, via cleavage by other MMPs or proteases, such as furin, urokinase, plasmin, and trypsin, or by treatment with organomercurial comounds. This preparation is highly purified native MMP-9 from stimulated human neutrophils. Requires activation prior to use.
Monomeric, native MMP-9 from stimulated human neutrophils. Requires activation prior to use.
Note: 1 mU = 1 milliunit.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol 2,4-DNP-Pro-Gln-Gly-Ile-Ala-Gly-Gln-D-Arg-OH per min at 37°C, pH 7.0.

Physical form

In 200 mM NaCl, 50 mM Tris-HCl, 5 mM CaCl₂, 1 µM ZnCl₂, 0.05% BRIJ® 35 Detergent, 0.05% NaN₃, pH 7.0.

Preparation Note

Prepared from stimulated neutrophils that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Reynolds, M. A., et al. 2003. Clin. Chem.49, 1733.
Kolkenbrock, H., et al. 1996. Biol. Chem. 377, 529.
Marcy, A.I., et al. 1991. Biochemistry30, 6476.
Stricklin, et al. 1983. Biochemistry22, 61.

Legal Information

Brij is a registered trademark of Croda International PLC
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service