Skip to Content
Merck
All Photos(1)

Documents

407313-M

Millipore

Anti-Integrin αIIb Mouse mAb (283.16B7)

liquid, clone 283.16B7, Calbiochem®

Synonym(s):

Anti-CD41

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

283.16B7, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITGA2B(3674)

General description

Mouse monoclonal antibody supplied as undiluted ascites. Recognizes the αIIb integrin chain.
Recognizes the αIIb integrin chain.
  • Antibody Target Gene Symbol: ITGA2B
  • Target Synonym: AI172977, ALPHA 2B, alphaIIb, CD41, CD41A, CD41B, GP2B, GPIIB, GTA, HPA3, INTEGRIN ALPHA 2B, Itga2b, LOC685269
  • Entrez Gene Name: integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41)
  • Hu Entrez ID: 16399
  • Rat Entrez ID:
  • This Anti-Integrin αIIb Mouse mAb (283.16B7) is validated for use in ELISA, Flow Cytometry, Paraffin Sections for the detection of Integrin αIIb.

    Immunogen

    Human
    integrin αIIb from M21-L-llb melanoma cells

    Application

    ELISA (see comments)

    Flow Cytometry (1:50-1:100)

    Paraffin Sections (1:50-1:100)

    Warning

    Toxicity: Standard Handling (A)

    Physical form

    Undiluted ascites.

    Reconstitution

    Following initial thaw, aliquot and freeze (-20°C).

    Other Notes

    Hynes, R.O. 1992. Cell.69, 11.
    Phillips, D.R., et al. 1991. Cell.65, 359.
    Albelda, S.M. and Buck, C.A. 1990. FASEB J.4, 2868.
    Buck, C.A. and Horwitz, A.F. 1987. Ann. Rev. Cell Biol.3, 179.
    Hynes, R.O. 1987. Cell.45, 549.
    Variables associated with assay conditions will dictate the proper working dilution.

    Legal Information

    CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

    Not finding the right product?  

    Try our Product Selector Tool.

    Storage Class Code

    10 - Combustible liquids

    WGK

    WGK 2

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service