Skip to Content
Merck
All Photos(1)

Key Documents

33-054

Sigma-Aldrich

PIP4-Kinase HTRF Assay

The PIP4 Kinase HTRF Assay kit provides a method for assaying PI(5)P-4 kinases in a homogenous 384 well format. Suitable for screening of potential inhibitors of these enzymes.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

species reactivity

human, mouse

manufacturer/tradename

HTRF®
Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

detection method

fluorometric (HTRF)

shipped in

dry ice

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Oncology
The PIP4 Kinase HTRF Assay kit provides a method for assaying PI(5)P-4 kinases in a homogenous 384 well format. Suitable for screening of potential inhibitors of these enzymes.
Used to detect/quantify: PIP4 Kinase

Biochem/physiol Actions

Target Sub-Family: PI3K/PI4K

Packaging

Kit capacity: 384 wells

Components

Reaction Buffer (4X)

DM A Detection Reagent

DM C Detection Reagent

DM E Detection Reagent

Phosphatidylinositol 5-phosphate (1mM)

Phosphatidylserine (1mM)

Stop Reagent D

Storage and Stability

12 months at -70°C

Legal Information

HTRF is a registered trademark of Cis Bio International
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service