Skip to Content
Merck
All Photos(2)

Key Documents

23-049-M

Sigma-Aldrich

p38/JTV-1 Protein, 10 µg

Full length, recombinant human p38/JTV-1. N-terminal cMyc, GST-tagged. For use in Enzyme Assays. Functions as a substrate in ubiquitination assays.

Synonym(s):

Aminoacyl tRNA synthase complex-interacting multifunctional protein 2, AIMP2, Multisynthase complex auxiliary component p38

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
₪3,528.00

₪3,528.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
₪3,528.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

₪3,528.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

100
300

recombinant

expressed in Sf21 cells

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

GenBank accession no.

General description

p38/JTV-1 is a scaffold required for the assembly and stability of the multi-tRNA synthetase complex, an essential enzyme that ligates specific amino acids to tRNAs before protein synthesis. It plays a pivotal role in the control of cell proliferation and has been shown to interact with FUSE binding protein, stimulating its ubquitination and degradation and leading to the down-regulation of the transcription factor c-Myc which is required for differentiation of functional alveolar type II cells. Genetic disruption of p38/JTV-1 in mice has been observed to cause neonatal lethality.

p38/JTV-1 is ubiquitinated by the E3 ligase Parkin, leading to its proteosomal degradation. It has been shown to accumulate in Parkin-deficient mice and in the brain tissue of patients with hereditary Parkinson’s, suggesting that it may play a role in the pathogenicity of this disease.

Application

Ubiquitination Cascade Comoponent: Substrate

Physical form

Purified using immobilized metal affinity chromatography.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service