Skip to Content
Merck
All Photos(1)

Key Documents

1.46602

Millipore

Fluid A Washing Solution

bottle volume 300 mL , filling volume

Synonym(s):

Fluid A for Sterility Testing, Rinsing Solution Fluid A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

Agency

EP 2.6.13
JP 4.06
USP 71

Quality Level

form

liquid

feature

closure type screw cap with septum

packaging

pack of 6 units
bottle of

technique(s)

sterile filtration: suitable

bottle capacity

500 mL

bottle volume

300 mL , filling volume

pH range

6.9-7.3

application(s)

pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for bacteria

General description

Fluid A is designed as a washing solution for sterility testing of aqueous solutions or water-soluble pharmaceutical products using membrane filtration method.
Fluid A washing solution in general serves as a rinse buffer and can be used for an initial rinse of the filter membrane to inactivate any inhibitors, if present.

Application

Fluid A washing solution is used as a general rinse buffer and is compatible with most test samples. It can also be used for dissolving or diluting the test samples, reconstituting commercial microbes or as transport medium for a broad range of microorganisms.

Analysis Note

The appearance of the medium is clear and colorless to slightly yellowish.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service