14-577-M
Hck Protein, active, 10 µg
Active, N-terminal His6-tagged, recombinant, human Hck, amino acids 230-497, for use in Kinase Assays.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
Quality Level
100
300
recombinant
expressed in baculovirus infected Sf21 cells
mol wt
Mw 34.1 kDa
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (kinase)
NCBI accession no.
UniProt accession no.
General description
N-terminal His6-tagged, recombinant, human Hck, amino acids 230-497
Product Source: Expressed by baculovirus in Sf21 insect cells
Application
Research Category
Inflammation & Immunology
Inflammation & Immunology
Biochem/physiol Actions
Protein Target: Hck
Target Sub-Family: TK
Packaging
Also available in 250 µg size --call for pricing and availability and reference catalog number 14-577M when ordering the 250 µg size.
Quality
Routinely evaluated by phosphorylation of Src substrate peptide
Physical form
Ni2+/NTA-agarose
Storage and Stability
6 months at -70°C
Other Notes
For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service