Skip to Content
Merck
All Photos(2)

Documents

1.37098

SAFC

Imidazole EMPROVE® EXPERT ACS

Pharma Manufacturing

Synonym(s):

Imidazole, Glyoxaline, Iminazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
MDL number:
UNSPSC Code:
47101613
EC Index Number:
206-019-2

grade

ACS reagent

Quality Level

vapor pressure

0.003 hPa ( 20 °C)

product line

EMPROVE® EXPERT

Assay

≥99% (perchloric acid titration)

form

solid

autoignition temp.

480 °C

technique(s)

API processing | cocrystal formation: suitable
API processing | salt formation: suitable

pH

10.5 (20 °C, 67 g/L in H2O)

bp

256 °C/1013 hPa

mp

90 °C

transition temp

flash point 145 °C

solubility

633 g/L

bulk density

500‑600 kg/m3

storage temp.

15-25°C

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service