Skip to Content
Merck
All Photos(1)

Documents

1.14798

Supelco

Cyanide Test Kit, colorimetric

0.03-0.07-0.13-0.2-0.3-0.5-1-2-5 mg/l CN⁻ MQuant®

Synonym(s):

Cyanide Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Cyanide Test, Method: colorimetric with color-disk comparator 0.03-0.07-0.13-0.2-0.3-0.5-1-2-5 mg/l CN⁻ MColortest®

Quality Level

specific analyte(s)

cyanide

CN- measuring range

0.03 -5 mg/L

compatibility

for use with MCOLORTEST

detection method

colorimetric

storage temp.

10-30°C

General description

The Cyanide test measures cyanide ions e.g. in drinking water, mineral water, wastewater, groundwater and surface water. The test kit contains the colour comparator for the visual measurement.
The MQuant® Cyanide test (formally MColortest®) measures free cyanide ions e.g. in
  • Groundwater and surface water
  • Drinking water and mineral water
  • Waters from aquaculture
  • Electroplating baths and bath solutions, electroplating wastewater
  • Industrial water
  • Wastewater and percolating water
This test is not suited for seawater.

Method
Cyanide ions react with a chlorinating agent to form cyanogen chloride, which in turn reacts with 1,3-dimethylbarbituric acid in the presence of pyridine to form a violet dye (König reaction). The cyanide concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation.
The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.
Using the refill pack 1.18457 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.

Warning

Toxicity: Standard Handling (A)

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service