Skip to Content
Merck
All Photos(1)

Documents

1.11724

Supelco

Spectromelt® A 14

51% di-lithium tetraborate/ 27% lanthane(III) oxide/ 12% lithium metaborate/ 10% lithium fluoride

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352402
NACRES:
NA.21

Quality Level

bulk density

1000‑1200 kg/m3

storage temp.

2-30°C

Analysis Note

Chloride (Cl): ≤ 50 ppm
Phosphate (PO₄): ≤ 20 ppm
Silicate (SiO₂): ≤ 300 ppm
Sulfate (SO₄): ≤ 50 ppm
Ag (Silver): ≤ 5 ppm
Al (Aluminium): ≤ 50 ppm
Ba (Barium): ≤ 5 ppm
Ca (Calcium): ≤ 50 ppm
Cd (Cadmium): ≤ 5 ppm
Co (Cobalt): ≤ 5 ppm
Cr (Chromium): ≤ 5 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 10 ppm
K (Potassium): ≤ 100 ppm
Mg (Magnesium): ≤ 10 ppm
Mn (Manganese): ≤ 5 ppm
Na (Sodium): ≤ 50 ppm
Ni (Nickel): ≤ 5 ppm
Pb (Lead): ≤ 20 ppm
Sr (Strontium): ≤ 5 ppm
Zn (Zinc): ≤ 10 ppm
Loss on ignition (800 °C): ≤ 1.0 %
Bulk density: about 110

Legal Information

SPECTROMELT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service