Skip to Content
Merck
All Photos(2)

Documents

1.07561

Supelco

Extran® AP 22

greener alternative

liquid, acidic concentrate (contains citric acid)

Synonym(s):

TM=["EXTRAN"] AP 22 cleaning agent; TM=["EXTRAN"] AP 22 detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103206
NACRES:
NA.21

Quality Level

form

liquid

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

32-35% Free acid (as citric acid)

pH

3.0 (20 °C, 2 g/L in H2O)

density

1.15 g/cm3 at 20 °C

greener alternative category

storage temp.

no temp limit

General description

Extran® AP 22 is an acidic laboratory cleaning agent. It cleans reliably and prevents residues from being carried over to the subsequent analysis, and ensuring accurate results.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is environmentally friendly having bio-degradable active ingredients. Click here for more information.

Application

Extran® AP 22 can be used both as a pre-wash agent and a rinsing agent with a neutralizing effect.
When used as:
  • A pre-wash agent, it primarily dissolves carbonates and hydroxides from the residues. Protein substances and organic bases, such as amines, are often removed.
  • A rinsing agent, it is especially suitable for removing remaining traces of alkali on the cleaned material or, in the case of solution carry-over, for neutralization.

This acidic cleaning agent is also well suited to the removal of calcareous deposits in the washing machine. The product is recommended for cases in which gentle conditions must be maintained. Particularly suitable for the gentle removal of calcareous deposits, e.g., on taps or sensitive metal and glass surfaces.

Analysis Note

Free acid (as citric acid): 32.0 - 35.0 %
Density (d 20 °C/ 4 °C): 1.140 - 1.150

Legal Information

EXTRAN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service