Skip to Content
Merck
All Photos(1)

Documents

1.07419

Millipore

Raffinose pentahydrate

for biochemistry

Synonym(s):

Raffinose pentahydrate, Melitose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
594.52
MDL number:
UNSPSC Code:
12352201
EC Index Number:
208-146-9

form

solid

pH

5.5-7.0 (25 °C, 100 g/L in H2O)

mp

80 °C

density

1.465 g/cm3 at 20 °C

storage temp.

15-25°C

InChI

1S/C18H32O16/c19-1-5-8(22)11(25)13(27)16(31-5)30-3-7-9(23)12(26)14(28)17(32-7)34-18(4-21)15(29)10(24)6(2-20)33-18/h5-17,19-29H,1-4H2/t5-,6-,7-,8+,9-,10-,11+,12+,13-,14-,15+,16+,17-,18+/m1/s1

InChI key

MUPFEKGTMRGPLJ-ZQSKZDJDSA-N

General description

for biochemistry

Application

Raffinose pentahydrate for biochemistry. CAS 17629-30-0, pH 5.5 - 7.0 (100 g/l, H₂O, 25 °C).

Analysis Note

Purity (HPLC)≥ 97.0 %Identity (IR-spectrum)passes testAppearancepasses testAppearance of solution (100 g/l, water)passes testSpec. rotation (α 20/D, 100 g/l, water, calc. on anhydrous substance)122.0 - 124.0 °Heavy metals (as Pb)≤ 0.001 %Reducing sugars (as glucose)≤ 1.0 %Water (according to Karl Fischer)14.0 - 16.0 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service