Skip to Content
Merck
All Photos(1)

Documents

1.05124

Supelco

Potassium thiocyanate

EMPLURA®

Synonym(s):

Potassium thiocyanate, Potassium sulfocyanide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CKNS
CAS Number:
Molecular Weight:
97.18
MDL number:
UNSPSC Code:
12352302
EC Index Number:
206-370-1
NACRES:
NA.21

vapor pressure

<0.001 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥98.0% (silver nitrate titration)

form

crystalline

potency

854 mg/kg LD50, oral (Rat)

pH

5.3-8.5 (20 °C, 50 g/L in H2O)

bp

>400 °C/1013 hPa (decomposition)

mp

177 °C

solubility

>1000 g/L

density

1.91 g/cm3 at 20 °C

bulk density

750‑1000 kg/m3

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.05%

cation traces

Fe: ≤0.0005%
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

InChI

1S/CHNS.K/c2-1-3;/h3H;/q;+1/p-1

InChI key

ZNNZYHKDIALBAK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Characteristics of Methyl Cellulose Based Solid Polymer Electrolyte Inserted with Potassium Thiocyanate as K(+) Cation Provider: Structural and Electrical Studies.: This research examines the role of potassium thiocyanate in enhancing the ionic conductivity of methyl cellulose-based solid polymer electrolytes. The findings contribute to advancements in materials science and battery technology (Aziz et al., 2022).


Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity: passes test
Chloride (Cl): ≤ 0.01 %
Sulfate (SO₄): ≤ 0.05 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service