Skip to Content
Merck

Skip To

1.02491

Chemizorb® H⁺

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


pH

9.0-12.0 (50 g/L in H2O, aqueous suspension)

Quality Segment

mp

1093 °C

General description

Absorbent and neutralizer for spilled acids, with indicator

Preparation Note

Store at +5°C to +30°C.

Legal Information

CHEMIZORB is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Chemizorb® H⁺


Storage Class

11 - Combustible Solids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters