Skip to Content
Merck
All Photos(2)

Key Documents

07-1519

Sigma-Aldrich

Anti-phospho Grb10 (Ser503) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Tumor necrosis factor ligand superfamily member 4, OX40 ligand, OX40L, CD252

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

primate (based on 100% sequence homology), porcine (based on 100% sequence homology), human (immunogen homology)

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer503)

Gene Information

human ... GRB10(2887)
mouse ... Grb10(14783)
pig ... Grb10(100188977)

General description

Grb10 (insulin receptor-binding protein Grb-IR) is a ubiquitous adapter protein which is expressed in both fetal and adult tissues. It is a multidomain protein that interacts with a wide array of cellular targets; the PH domain allows interaction with several subtypes of phosphoinositides; the Ras-associating domain binds a wide variety of Ras GTPases; and the SH domain facilitates binding to membrane-bound tyrosine kinase receptors which link Grb10 to different signaling pathways. Most notably, Grb10 associates with the insulin receptor at phosphotyrosine residues via its C-terminal BPS (between PH and SH) domain and inhibits the enzymatic activity of the insulin receptor. Grb10 also acts downstream of the insulin receptor pathway to inhibit activation of AKT-1. In addition, Grb10 suppresses Wnt signalling by binding to the low-density lipoprotein receptor. In fetal tissues, Grb10 may inhibit growth and placental size.

Specificity

Other homologies: Rat (91% sequence homology).
This antibody recognizes Grb10 phosphorylated at (Ser503).

Immunogen

Epitope: Phosphorylated (Ser503)
KLH-conjugated linear peptide corresponding to human GRB10 phosphorylated at (Ser503).

Application

Peptide Inhibition Assay Analysis: 1 µg/mL from a representative lot detected phospho-Grb10 (Ser503) on 10 µg of embryonic fibroblast cell lysate + non-phospho-Grb10 (Ser503) peptide. However, the antibody was inhibited when incubated with a phospho-Grb10 (Ser503) peptide.
Research Category
Signaling
Research Sub Category
Developmental Signaling
This Anti-phospho Grb10 (Ser503) Antibody is validated for use in Peptide Inhibition Assay, Western Blotting for the detection of phospho Grb10 (Ser503).

Quality

Evaluated by Western Blot in mouse embryonic fibroblast cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected phospho-Grb10 (Ser503) on 10 µg of mouse embryonic fibroblast cell lysate.

Target description

~75 kDa observed. An uncharacterized band appears at ~16 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse embryonic fibroblast cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena M Lucchetta et al.
Cell stem cell, 20(5), 609-620 (2017-03-28)
Organ fitness depends on appropriate maintenance of stem cell populations, and aberrations in functional stem cell numbers are associated with malignancies and aging. Symmetrical division is the best characterized mechanism of stem cell replacement, but other mechanisms could also be
Katarzyna M Tyc et al.
Developmental cell, 42(3), 256-270 (2017-08-09)
Proper regulation of the germline transcriptome is essential for fertility. In C. elegans, germline homeostasis hinges on a complex repertoire of both silencing and activating small RNA pathways, along with RNA processing. However, our understanding of how fundamental RNA processing steps

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service