Skip to Content
Merck
All Photos(1)

Documents

CH4428741590

ChemBridge Corporation

N,N-dimethyl-2-(4-piperidinyloxy)ethanamine dihydrochloride heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
371.3
MDL number:

Iupac Name

N,N-dimethyl-2-(piperidin-4-yloxy)ethan-1-amine dihydrochloride heptahydrate

Inchi Code

1S/C9H20N2O.2ClH.7H2O/c1-11(2)7-8-12-9-3-5-10-6-4-9;;;;;;;;;/h9-10H,3-8H2,1-2H3;2*1H;7*1H2

InChI key

KNIVNHFLJHEWCL-UHFFFAOYSA-N

Salt Data

2HCl 7H2O

Purity

95%

Country of Origin

KZ

Product Link

https://www.hit2lead.com/building-blocks/4002975

MSDS

https://www.hit2lead.com/sds/4002975_sds.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service