Skip to Content
Merck
All Photos(1)

Key Documents

880710C

Avanti

14:0 PEG1000 PE

Avanti Research - A Croda Brand 880710C

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-1000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (880710C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880710C

concentration

10 mg/mL (880710C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O.[NH4+]

Application

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-1000] (14:0 PEG1000 PE) is suitable for use in lipid aggregates to characterize and study its use as a coating and a carrier in capillary electrophoresis.[1] It might be used to prepare sterically stabilized liposome and study its characteristics by small-angle X-ray scattering (SAXS) method.[2]

Biochem/physiol Actions

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-1000] (14:0 PEG1000 PE) is useful in preparing polyethylene glycol (PEG)-stabilized lipid aggregates, which serve as model membranes in biotechnical and pharmaceutical applications. PEG conjugated lipids improve liposome packing in the water/hydrocarbon interface. PEGs of molecular mass less than or equal to 5000 is preferred for vesicle coating serving for efficient delivery of the drug.[1]

Packaging

5 mL Clear Glass Sealed Ampule (880710C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria V Lindén et al.
Electrophoresis, 29(4), 852-862 (2008-01-24)
PEG-stabilized lipid aggregates are a promising new class of model membranes in biotechnical and pharmaceutical applications. CE techniques, field-flow fractionation, light scattering, quartz crystal microbalance (QCM), and microscopic techniques were used to study aggregates composed of 1-palmitoyl-2-oleyl-sn-glycero-phosphatidylcholine (POPC) and PEG-lipid
Z Varga et al.
Chemistry and physics of lipids, 165(4), 387-392 (2012-01-10)
Synchrotron small-angle X-ray scattering analysis of the bilayer structure of a pharmacologically relevant sterically stabilized liposome system is presented. Describing the electron density profile of the bilayer with the superposition of Gaussian functions, the contribution of the poly(ethylene glycol) (PEG)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service