Skip to Content
Merck
All Photos(1)

Documents

870336P

Avanti

Soy PS

Avanti Research - A Croda Brand

Synonym(s):

L-α-phosphatidylserine (Soy, 99%) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 2 × 100 mg (870336P-200mg)
pkg of 5 × 100 mg (870336P-500mg)
pkg of 1 × 25 mg (870336P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C40H74NO10P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-39(43)51-36(34-49-52(46,47)50-35-37(41)40(44)45)33-48-38(42)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h11,13,17-18,36-37H,3-10,12,14-16,19-35,41H2,1-2H3,(H,44,45)(H,46,47);/q;+1/p-1/b13-11-,18-17-;/t36-,37+;/m1./s1

InChI key

WPLLCKLCLMRAPV-WHVRKWPVSA-M

General description

Soy-PS is a phosphotidylserine. It is enzymatically obtained from soybean lecithin along with L-serine.

Application

Soy PS has been used in the synthesis of multilamellar vesicles. It has also been used as a standard to identify spots in thin layer chromatography.

Packaging

5 mL Clear Glass Sealed Ampule (870336P-200mg)
5 mL Clear Glass Sealed Ampule (870336P-25mg)
5 mL Clear Glass Sealed Ampule (870336P-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

CLD1 Reverses the Ubiquinone Insufficiency of Mutant cat5/coq7 in a Saccharomyces cerevisiae Model System
Kar A, et al.
Testing, 11(9) (2016)
CLD1 Reverses the Ubiquinone Insufficiency of Mutant cat5/coq7 in a Saccharomyces cerevisiae Model System
Kar A, et al.
PLoS ONE, 11(9) (2016)
Soybean-derived phosphatidylserine improves memory function of the elderly Japanese subjects with memory complaints
Kato-Kataoka A, et al.
Journal of Clinical Biochemistry and Nutrition (2010)
The catalytic domains of Clostridium sordellii lethal toxin and related large clostridial glucosylating toxins specifically recognize the negatively charged phospholipids phosphatidylserine and phosphatidic acid
Varela Chavez C, et al.
Cellular Microbiology, 17(10), 1477-1493 (2015)
Yuki Nakamura et al.
Nature communications, 5, 3553-3553 (2014-04-05)
Arabidopsis FT protein is a component of florigen, which transmits photoperiodic flowering signals from leaf companion cells to the shoot apex. Here, we show that FT specifically binds phosphatidylcholine (PC) in vitro. A transgenic approach to increase PC levels in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service