852465P
Avanti
C18(Plasm) LPC
1-O-1′-(Z)-octadecenyl-2-hydroxy-sn-glycero-3-phosphocholine, powder
Synonym(s):
PC(18:1p/0:0); 110694
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 10 mg (852465P-10mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 852465P
lipid type
phospholipids
cardiolipins
shipped in
dry ice
storage temp.
−20°C
General description
C18(Plasm) LPC is a plasmalogen.
Application
C18(Plasm) LPC has been used to test the lysoplasmalogenase activity of YhhN proteins.
Packaging
5 mL Amber Glass Screw Cap Vial (852465P-10mg)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The YhhN protein of Legionella pneumophila is a Lysoplasmalogenase
Biochimica et Biophysica Acta - Biomembranes, 1848(2), 742-751 (2015)
Biochimica et biophysica acta, 1848(2), 742-751 (2014-12-03)
Lysoplasmalogenase catalyzes hydrolytic cleavage of the vinyl-ether bond of lysoplasmalogen to yield fatty aldehyde and glycerophospho-ethanolamine or glycerophospho-choline. We recently purified lysoplasmalogenase from rat liver microsomes and identified the protein as TMEM86B, an integral membrane protein that is a member
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service