Key Documents
Z684821
Ace cylindrical reaction flask
capacity 2 L, O-ring grooved flange, valve size 0-20 mm , zero dead space
Select a Size
1 EA
₪5,382.00
₪5,382.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
₪5,382.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
₪5,382.00
Please contact Customer Service for Availability
material
borosilicate glass
cylindrical
single-neck
sterility
non-sterile
feature
cap: no
packaging
case of 1
manufacturer/tradename
Ace Glass 6518-14
capacity
2 L
valve size
0-20 mm , zero dead space
General description
Heavy wall cylindrical reaction flask with Schott grooved flange opening. Flask has Zero Dead Space, (ZDS), bottom valve. The valve features no material hang-up in the fully open position. When closed, the valve seats flush with the bottom of the flask. Includes one CAPFE O-ring. See Ace 6517 (Z508233) for quick release clamps.
Choose from one of the most recent versions:
Certificates of Analysis (COA)