Z681164
Colored Microscope slides
blue
Synonym(s):
235502202
About This Item
Recommended Products
material
soda-lime glass
manufacturer/tradename
DURAN® 235502202
W × L × H
26 mm × 76 mm × 1.0 mm
color
blue
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
These 90° ground white PRINT microscope slides with rounded corners are ideal for use in automatic machines, as glass breakages in the cassettes and on the conveyer belts of the machines can then be avoided.
- Microscope slide in standard dimensions 26 x 76 mm
- Glass thickness 1.0 ± 0.05 mm
- The edges of the glass slides are uniform and crystal clear
- Very uniform wettability
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service