Documents
Z680508
Duran® GL 45 connection system caps and accessories
spare membrane filter for pressure equalization set, 0.2 μm
Synonym(s):
1129819
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
blue-gray polypropylene
manufacturer/tradename
DURAN® 1129819
parameter
-40-140 °C temp. limit
General description
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Features and Benefits
• Manufactured from PP, PTFE, silicone• Accessories designed for use with the DURAN® GL 45 2/3 port tubing connection caps or GL 45 / GLS 80® Stirred Bottle Reactor caps• Safe and secure liquid transfer into or out of laboratory bottles• Four inserts are suitable for rigid or semi-rigid tubing with a range of outer diameters: 1.6, 3.0, 3.2 or 6.0 mm• Open-topped blue PP GL 14 pass-through tubing cap for use with the inserts• Closed-top red PBT GL 14 cap for sealing unused ports• Sterile pressure equilisation is possible with pressure compensation set
Legal Information
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group