Documents
Z568740
Simriz® high-performance O-ring for QianCap
15-S
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
manufacturer/tradename
Q Labtech
General description
Have excellent compatibility with most of the chemicals, including inorganic and organic acids and bases, alcohols, aliphatic and chlorinated hydrocarbons, aromatics, ketones, esters, ethers and aldehydes and fuel oils.
Legal Information
QianCap is a trademark of Q Labtech LLC
Simriz is a registered trademark of Carl Freudenburg KG