Z267562
Round crystal window
CaF2, diam. × thickness 25 mm × 2 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
CaF2
technique(s)
IR spectroscopy: suitable
diam. × thickness
25 mm × 2 mm
transmittance
77,000-111,000 (wavenumbers (cm-1))
refractive index
n/D 1.42 (at 2 μm)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
A strong crystal; resists most acids and alkalis; withstands high-pressure; insoluble in water; no fog.
Cut and polished quality crystal windows.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service