Z218758
Ace giant Soxhlet extraction apparatus
flask capacity 1 L, extractor capacity 0.5 L, thimble I.D. × L 55 mm × 165 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
size
A
manufacturer/tradename
Ace Glass 681010
Overall H
39 in.
extractor capacity
0.5 L
flask capacity
1 L
thimble I.D. × L
55 mm × 165 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Complete with bulb type condenser and one flask. Cycling rate may be doubled over conventional extractors. All joints are interchangeable. Size C is supplied with adapter Z218871 for connecting the XL extractor joint and condenser. Order giant thimbles separately.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service