Skip to Content
Merck
All Photos(1)

Documents

W530345

Sigma-Aldrich

Buchu leaf oil, Betulina

natural, FG

Synonym(s):

Agathosma betulina

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2169
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Barosma betulina

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 173.65 (172.51)

form

liquid

composition

Limonene, 16.6%
Linalol, 0.5%
Pulegone, 35-45%

refractive index

n20/D 1.475

density

0.922 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; green; tropical; herbaceous; minty; fruity

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.0 °F - closed cup

Flash Point(C)

67.78 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service