Documents
SYNV131003
Synthware conical bottom reaction vial with holed compression cap
3 mL, joint: ST/NS 14/10
Synonym(s):
Synthware V131003, reaction tube
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS14/10
packaging
pkg of 1 ea
manufacturer/tradename
Synthware V131003
capacity
3 mL
Related Categories
General description
Supplied with one holed compression cap, one Viton® O-ring and one PTFE -faced silicone rubber septum. For replacement caps, see RC112; For Viton® O-rings, see R014; for septa, see R1420.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company