SLW1200/04M
Pyrex® Kjeldahl flask
plain neck
Synonym(s):
Pyrex, SciLabware 1200/04M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
Pyrex flask
round bottom
single-neck
feature
plain neck
packaging
pack of 10 ea
manufacturer/tradename
1200/04M
SciLabware
H
240 mm
bulb diam.
58 mm
capacity
100 mL
100 mL
neck I.D. × O.D.
17 mm × 22 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Suitable for the determination of nitrogen etc.
- Manufactured from Pyrex borosilicate glass
- Withstands high temperatures
- Resistant to chemical corrosion
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service