Skip to Content
Merck
All Photos(1)

Documents

QBD10501

Sigma-Aldrich

Azido-dPEG®4-NHS ester

Synonym(s):

3-(Azidotetra(ethyleneoxy))propionic acid succinimidyl ester, NHS-Peg4-Azide, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N4O8
CAS Number:
Molecular Weight:
388.37
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent
reaction type: click chemistry

functional group

NHS ester
azide

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(ON1C(CCC1=O)=O)CCOCCOCCOCCOCCN=[N+]=[N-]

InChI

1S/C15H24N4O8/c16-18-17-4-6-24-8-10-26-12-11-25-9-7-23-5-3-15(22)27-19-13(20)1-2-14(19)21/h1-12H2

InChI key

OIGKWPIMJCPGGD-UHFFFAOYSA-N

Features and Benefits

The azido-dPEG®4-NHS ester contains an azide function on one end of a single molecular weight dPEG® spacer (17.7 Å) and a reactive group on the other end of the spacer. The dPEG® spacer is hydrophilic and non-immunogenic and improves the water solubility of the target molecule while reducing the immunogenicity and increasing the hydrodynamic volume of the target. NHS ester readily reacts with amines in aqueous solution or organic solvent. The azide group reacts with an alkyne in the well-known click chemistry reaction. The click chemistry reaction proceeds by copper(I) or ruthenium catalysis or in a strain-catalyzed reaction with certain types of alkyne partners.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service