MT51344763
Mettler Toledo Ionic Strength Adjuster
Nitrate, perfectION™ ISE combination electrodes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
manufacturer/tradename
Mettler-Toledo 51344763
volume
475 mL
Looking for similar products? Visit Product Comparison Guide
General description
Ionic strength adjuster (ISA) provides a constant background ionic strength for samples and standards.
Legal Information
perfectION is a trademark of Mettler-Toledo, Inc.
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service