Skip to Content
Merck
All Photos(1)

Documents

H60255

Sigma-Aldrich

Dopamine hydrochloride

98%

Synonym(s):

2-(3,4-Dihydroxyphenyl)ethylamine hydrochloride, 3,4-Dihydroxyphenethylamine hydrochloride, 3-Hydroxytyramine hydrochloride, 4-(2-Aminoethyl)-1,2-benzenediol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2CH2NH2·HCl
CAS Number:
Molecular Weight:
189.64
Beilstein:
3656720
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

mp

248-250 °C (lit.)

SMILES string

Cl[H].NCCc1ccc(O)c(O)c1

InChI

1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H

InChI key

CTENFNNZBMHDDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florian Ponzio et al.
Journal of colloid and interface science, 459, 29-35 (2015-08-12)
Polydopamine (PDA) coatings appear as a universal functionalization methodology allowing to coat the surface of almost all kinds of known materials with a conformal, stable, robust and reactive material. Relatively few investigations were dedicated to the incorporation of other molecules
Alia H Clark et al.
Bioorganic & medicinal chemistry, 19(18), 5420-5431 (2011-08-25)
To assess the effect of conformational mobility on receptor activity, the β-phenyl substituent of dopamine D(1) agonist ligands of the phenylbenzazepine class, (±)-6,6a,7,8,9,13b-hexahydro-5H-benzo[d]naphtho[2,1-b]azepine-11,12-diol (8), and its oxygen and sulfur bioisosteres 9 and 10, respectively, were synthesized as conformationally-restricted analogs of
Shuang Chang et al.
Nature chemical biology, 4(4), 256-263 (2008-03-11)
Fragile X syndrome is caused by the functional loss of the fragile X mental retardation 1 (FMR1) gene. Deletion of the FMR1 ortholog in Drosophila melanogaster (Fmr1) recapitulates many phenotypes associated with fragile X syndrome. We have discovered that Fmr1
Jing Yuan et al.
Nature chemical biology, 5(10), 765-771 (2009-09-08)
Studies of gene function and molecular mechanisms in Plasmodium falciparum are hampered by difficulties in characterizing and measuring phenotypic differences between individual parasites. We screened seven parasite lines for differences in responses to 1,279 bioactive chemicals. Hundreds of compounds were
Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service