Skip to Content
Merck
All Photos(1)

Documents

GF05735134

Bismuth-lead alloy, Bi55.5Pb44.5

1000g lump, 100mm max. lump size

Synonym(s):

BI076100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141706
NACRES:
NA.23

form

lumps

manufacturer/tradename

Goodfellow 057-351-34

weight

1000 g

max. lump size

100 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service