Key Documents
DWKW242832-A
Wheaton® Dropping Bottle
white bottle, capacity 6 mL, case of 1000
Select a Size
1 PKG
₪5,137.00
₪5,137.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
₪5,137.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
₪5,137.00
Please contact Customer Service for Availability
material
white bottle
packaging
pkg of 1 packs
case of 1000
manufacturer/tradename
WHEATON® W242832-A
capacity
6 mL
Application
Opthalmic, IVD Kits, Other
Features and Benefits
  • Bottle made of natural or white LDPE (Low Density Polyethylene
  • White LDPE bottle helps protect UV light sensitive samples
  • Dropper tip caps and tips sold separately
  • Packaged in cases of 100 or 1000
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)