Skip to Content
Merck
All Photos(1)

Key Documents

DWKW242821

Wheaton® Dropping Bottle

natural bottle, capacity 3 mL

Synonym(s):

W242821

Sign Into View Organizational & Contract Pricing

Select a Size

100 EACH
₪641.00

₪641.00


Please contact Customer Service for Availability


Select a Size

Change View
100 EACH
₪641.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

₪641.00


Please contact Customer Service for Availability

material

natural bottle

packaging

pkg of 100 ea

manufacturer/tradename

WHEATON® W242821

capacity

3 mL

Looking for similar products? Visit Product Comparison Guide

Application

Opthalmic, IVD Kits, Other

Features and Benefits

  • Bottle made of natural or white LDPE (Low Density Polyethylene
  • White LDPE bottle helps protect UV light sensitive samples
  • Dropper tip caps and tips sold separately
  • Packaged in cases of 100 or 1000

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service