Skip to Content
Merck
All Photos(1)

Documents

D94003

Sigma-Aldrich

2-(Diethylamino)ethanol

99%

Synonym(s):

N,N-Diethylethanolamine, DEAE, DEEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCH2CH2OH
CAS Number:
Molecular Weight:
117.19
Beilstein:
741863
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

vapor density

4.04 (vs air)

vapor pressure

1 mmHg ( 20 °C)

Assay

99%

form

liquid

expl. lim.

11.7 %

refractive index

n20/D 1.441 (lit.)

bp

161 °C (lit.)

density

0.884 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CCO

InChI

1S/C6H15NO/c1-3-7(4-2)5-6-8/h8H,3-6H2,1-2H3

InChI key

BFSVOASYOCHEOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H P Lesch et al.
Gene therapy, 18(6), 531-538 (2011-01-21)
Lentivirus can be engineered to be a highly potent vector for gene therapy applications. However, generation of clinical grade vectors in enough quantities for therapeutic use is still troublesome and limits the preclinical and clinical experiments. As a first step
Prashanti B Konduru et al.
Environmental science & technology, 44(6), 2138-2143 (2010-02-16)
N,N-Diethylethanolamine (DEEA) is a very promising absorbent for CO(2) removal from gaseous streams, as it can be prepared from renewable resources. Aqueous mixtures of DEEA and piperazine (PZ) are attractive for the enhancement of CO(2) capture, due to the high
Jian-Bo Qu et al.
Journal of chromatography. A, 1216(37), 6511-6516 (2009-08-12)
Using agarose coated gigaporous polystyrene microspheres as a base support, a novel anion exchanger (DEAE-AP) has been developed after functionalization with diethylaminoethyl chloride. The gigaporous structure, static adsorption behavior, and chromatographic properties of DEAE-AP medium were characterized and compared with
Stephen P Miller et al.
Chembiochem : a European journal of chemical biology, 15(8), 1145-1153 (2014-05-07)
An active site lysine essential to catalysis in isocitrate dehydrogenase (IDH) is absent from related enzymes. As all family members catalyze the same oxidative β-decarboxylation at the (2R)-malate core common to their substrates, it seems odd that an amino acid
Elizabeth C Schramm et al.
Blood, 125(15), 2359-2369 (2015-01-23)
The pathogenesis of atypical hemolytic uremic syndrome (aHUS) is strongly linked to dysregulation of the alternative pathway of the complement system. Mutations in complement genes have been identified in about two-thirds of cases, with 5% to 15% being in C3.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service