Key Documents
BAB602102900
SP Bel-Art® H-B Frio-Temp® Calibrated Electronic Verification Thermometer
temp. range at -50-300 °C, Refrigerator Calibration
Synonym(s):
Calibrated refrigerator thermometer
About This Item
UNSPSC Code:
41112303
NACRES:
NB.71
material
glass bottle
stainless steel probe
feature
Alarm, Timer
Refrigerator Calibration
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B60210-2900
range of application measuring range
-50-300 °C, resolution: 1 °F/C (±1°C from -20/130°C; ±2°C from -50/-21°C and 131/200°C; otherwise ±4°C)
parameter
temp. range at -50-300 °C
H × W
18 mm × 52 mm
probe L
105 mm , 4.1"
battery power
AAA
General description
Digital thermometers verify sample temperatures in freezers, refrigerators, incubators and ovens. Available calibrated for specific applications and equipment
Note: Plastic bottles as indicated in table cannot be used in ovens. Glass bottle and cap included with products as indicated in the table should not be used in conditions where temperatures may exceed 200°C.
Features and Benefits
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
  • Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish) Individually serialized, CE marked
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
FRIO-Temp is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)